missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, 99.0 to 103.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Cantidad:
500 g
2.5 kg
Envase:
Poly Bottle
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Calcium Nitrate Tetrahydrate |
Punto de fusión | 44°C |
pH | 4 to 7 at 20°C |
Cantidad | 500 g |
Fórmula lineal | Ca(NO3)2 · 4H2O |
Número UN | NONH for all modes of transport |
Formula Weight (peso de la fórmula) | 236.15g/mol |
Porcentaje de pureza | 99.0 to 103.0% |
Grado | ACS Reagent |
Envase | Poly Bottle |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'