Learn More
Cadence Science™ Nonflexible Stainless Steel Animal Feeding Needles
Product Code. 16227951
Change view
Click to view available options
Gauge:
13G
Length (English):
3 in.
Diameter (Metric) Ball:
4 mm
Unit Size:
Pack of 12
Product Code. Gauge Length (English) Diameter (Metric) Ball Unit Size
16227951 13G 3 in. 4 mm Pack of 12
1 options
This item is not returnable. View return policy
Product Code. 16227951 Supplier Cadence Science™ Supplier No. 7924
Fecha estimada de envoi: 05-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
For feeding and dosing without injury to sensitive tissues.
Specifications
Product Type Animal Feeding Needle
Material Stainless Steel (304)
Disposable Reusable
For Use With (Application) Animal feeding and dosing without injury to sensitive tissues, ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals
Gauge 13G
Sterility Non-sterile
Length (English) 3 in.
Length (Metric) 7.62 cm
Diameter (Metric) Ball 4 mm