missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CACNB1 (Human) Recombinant Protein
Human CACNB1 full-length ORF ( NP_000714.3, 1 a.a. - 598 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000782-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MVQKTSMSRGPYPPSQEIPMEVFDPSPQGKYSKRKGRFKRSDGSTSSDTTSNSFVRQGSAESYTSRPSDSDVSLEEDREALRKEAERQALAQLEKAKTKPVAFAVRTNVGYNPSPGDEVPVQGVAITFEPKDFLHIKEKYNNDWWIGRLVKEGCEVGFIPSPVKLDSLRLLQEQKLRQNRLGSSKSGDNSSSSLGDVVTGTRRPTPPASAKQKQKSTEHVPPYDVVPSMRPIILVGPSLKGYEVTDMMQKALFDFLKHRFDGRISITRVTADISLAKRSVLNNPSKHIIIERSNTRSSLAEVQSEIERIFELARTLQLVALDADTINHPAQLSKTSLAPIIVYIKITSPKVLQRLIKSRGKSQSKHLNVQIAASEKLAQCPPEMFDIILDENQLEDACEHLAEYLEAYWKATHPPSSTPPNPLLNRTMATAALAASPAPVSNLQGPYLASGDQPLERATGEHASMHEYPGELGQPPGLYPSSHPPGRAGTLRALSRQDTFDADTPGSRNSAYTELGDSCVDMETDPSEGPGLGDPAGGGTPPARQGSWEDEEEDYEEELTDNRNRGRNKARYCAEGGGPVLGRNKNELEGWGRGVYIR
Especificaciones
NP_000714.3 | |
calcium channel, voltage-dependent, beta 1 subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CACNB1 | |
GST |
782 | |
Wheat germ expression system | |
10 μg | |
CAB1, CACNLB1, CCHLB1, MGC41896 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |