missing translation for 'onlineSavingsMsg'
Learn More

CA3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3C10A2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66608-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818645

  • 319.00€ / 150 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Carbonic anhydrase III (CAIII) is a member of carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal.
Especificaciones

Especificaciones

CA3
3C10A2
Unconjugated
CA3
Liquid
Ca3
CA3 Fusion Protein Ag7513
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Porcine, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07451, P14141, P16015, Q5S1S4
CA III, CA3, CAIII, Car3, Carbonate dehydratase III, Carbonic anhydrase 3, Carbonic anhydrase III
Mouse
IgG2b
150 μL
-20°C
Primary
12350, 494016, 54232, 761
Videos
SDS
Documentos
Certificados