Learn More
C9orf9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26034-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Product No. 26034-1-AP is a rabbit polyclonal antibody raised against the full-length of human C9orf9. Two isoforms were produced by alternative splicing with predicted MW of 19 and 26 kDa. Two bands at 16 kDa and 30 kDa could be detected using the presen
SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.Especificaciones
C9orf9 | |
Polyclonal | |
Unconjugated | |
SPACA9 | |
C9orf9, Uncharacterized protein C9orf9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11092, 296610, 69987 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TPM5, Q96E40 | |
Spaca9 | |
C9orf9 Fusion Protein Ag23320 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |