missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25994-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FAM198B is a protein coding gene.Especificaciones
C4orf18 | |
Polyclonal | |
Unconjugated | |
FAM198B | |
AD021, AD036, C4orf18, ENED, FAM198B, Protein FAM198B, UNQ2512/PRO6001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51313, 68659 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UPI1, Q6UWH4 | |
FAM198B | |
C4orf18 Fusion Protein Ag23312 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |