missing translation for 'onlineSavingsMsg'
Learn More

C4.4A/LYPD3 Antibody (213), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89995JF646

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213796

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

C4.4A/LYPD3 Monoclonal specifically detects C4.4A/LYPD3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

C4.4A/LYPD3
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Protein A purified
RUO
27076
Store at 4°C in the dark.
IgG
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
213
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4
This antibody was obtained from a rabbit immunized with purified, recombinant Human C4.4A/LYPD3 (NP_055215.2; Met 1-His 286).
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales