missing translation for 'onlineSavingsMsg'
Learn More

C20orf7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 26359-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814855

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones

Especificaciones

C20orf7
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5TEU4
C20orf7, NDUFAF5
Rabbit
IgG
150 μL
-20°C
Primary
79133
Western Blot
Unconjugated
NDUFAF5
Liquid
NDUFAF5
C20orf7 Fusion Protein Ag23822
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados