missing translation for 'onlineSavingsMsg'
Learn More

C16orf72 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87100

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213829

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

C16orf72 Polyclonal specifically detects C16orf72 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

C16orf72
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
29035
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149
The immunogen is a synthetic peptide directed towards the middle region of human C16orf72
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales