missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf93 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26395-1-AP-150UL
Additional Details : Peso : 0.00010kg
Beschreibung
C14orf93 is a protein coding gene.Spezifikation
C14orf93 | |
Polyclonal | |
Unconjugated | |
C14orf93 | |
C14orf93 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60686 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H972 | |
C14orf93 | |
C14orf93 Fusion Protein Ag23838 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |