missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf74 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25758-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The exact function of C11orf74 remains unknown.Especificaciones
C11orf74 | |
Polyclonal | |
Unconjugated | |
C11orf74 | |
C11orf74, Protein HEPIS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
119710 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VG3 | |
C11orf74 | |
C11orf74 Fusion Protein Ag22770 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |