Learn More
Thermo Scientific Chemicals C-Myc Peptide
For cellular and molecular biology applications
Marca: Thermo Scientific Chemicals J66846.#0
Detalles adicionales : Peso : 0.00005kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
HPLC | |
Solid | |
C-Myc Peptide | |
RUO - research use only |
-30°C to -10°C | |
1203.29 | |
5 mg | |
Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu |
RUO – Research Use Only