Learn More
C-Desmethyl Metoprolol, TRC
Product Code. 30364393
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364393 100 mg 100mg
30350902 10 mg 10mg
30354926 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364393 Supplier Toronto Research Chemicals Supplier No. TRCD292068100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109632-08-8
Molecular Formula C14 H23 N O3
Molecular Weight (g/mol) 253.34
Synonym (2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol, Metoprolol Succinate Imp. A (EP), Metoprolol Tartrate Imp. A (EP)
IUPAC Name 1-(ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
SMILES CCNCC(O)COc1ccc(CCOC)cc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H23NO3/c1-3-15-10-13(16)11-18-14-6-4-12(5-7-14)8-9-17-2/h4-7,13,15-16H,3,8-11H2,1-2H3
Formula Weight 253.1678
Chemical Name or Material (2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol