missing translation for 'onlineSavingsMsg'
Learn More

JP Selecta™ Bunsen Burners

Product Code. 12002445
Change view
Click to view available options
Gas Type:
Butane
Natural Gas
Height (Metric):
140 mm
165 mm
170 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gas Type Height (Metric) unitSize
12002445 Natural Gas 170 mm Each
12048363 Natural Gas 165 mm Each
12082435 Natural Gas 140 mm Each
12157691 Butane 140 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12002445 Supplier JP Selecta™ Supplier No. 7000661
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Bunsen Burner
Gas Type Natural Gas
Diameter (Metric) 11 mm
Height (Metric) 170 mm
Includes Gas and Air Regulator
Weight (Metric) 0.01 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.