Description
- High throughput due to advanced design of condensers
- Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
- Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
- Maximum stability and safety with plastic and glass coating
Specifications
Specifications
| Material | Glass |
| Type | Cold Trap Condenser |
| For Use With (Application) | Evaporator Condensation |
| For Use With (Equipment) | Rotavapor R-80 |
| Material (Coating) | Plastic and Glass |