Learn More
Bu-1a Antibody (21-1A4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP1-28321
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213690
  • 566.94€ / 0.25 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
Bu-1a Monoclonal specifically detects Bu-1a in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Especificaciones
Especificaciones
Bu-1a/b
Monoclonal
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Bursa cells from one-day-old H.B15 (Bu-1a/b) strain chickens
0.25 mg
Primary
Chicken
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry
21-1A4
Biotin
Mouse
Protein A or G purified
RUO
Chicken Bu-1a/b alloantigen, Mr 70 kDa
Store at 4C. Do not freeze.
IgG1 κ
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
For Research Use Only