missing translation for 'onlineSavingsMsg'
Learn More

BTBD6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-84523-0.1ML

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213839

  • 455.71€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

BTBD6 Polyclonal specifically detects BTBD6 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

BTBD6
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
90135
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
BDPLBTB domain protein BDPL, BTB (POZ) domain containing 6, BTB/POZ domain-containing protein 6, glucocorticoid receptor AF-1 coactivator-1, Lens BTB domain protein
The immunogen is a synthetic peptide directed towards the C-terminal region of Human BTBD6
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales