missing translation for 'onlineSavingsMsg'
Learn More

BRPF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87030

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215319

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

BRPF1 Polyclonal specifically detects BRPF1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

BRPF1
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
7862
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
bromodomain and PHD finger containing, 1, Bromodomain and PHD finger-containing protein 1, peregrin, Protein Br140
The immunogen is a synthetic peptide directed towards the middle region of human BRPF1
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales