missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sal sódica azul de bromofenol, solución acuosa al 0,04 % p/v
Click to view available options
Cantidad:
100 mL
500 mL
6 x 100 mL
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Bromophenol Blue sodium salt |
Concentración | 0.04% w/v aq. soln. |
Índice Merck | 141444 |
Formula Weight (peso de la fórmula) | 691.96 |
Color | Rojo |
Cantidad | 6 x 100 mL |
Forma física | Líquido |
RUO - Para uso exclusivo en investigación
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'