Learn More
Bromo-Peg3-Bromide, TRC
Product Code. 30332683
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30332683 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 30332683 Supplier Toronto Research Chemicals Supplier No. TRCB80054050MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bromo-Peg3-Bromide
CAS 31255-10-4
Molecular Formula C6H12O2Br2
Quantity 50 mg
Synonym Ethane, 1,2-Bis(2-Bromoethoxy)-
InChI Formula InChI=1S/C6H12Br2O2/c7-1-3-9-5-6-10-4-2-8/h1-6H2
SMILES BrCCOCCOCCBr
IUPAC Name 1,2-bis(2-bromoethoxy)ethane
Molecular Weight (g/mol) 275.96
Formula Weight 273.9204