Learn More
BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 24-well Standard Plate
Shop All Brand ProductsDescription
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Available with pureGrade™ S and cellGrade™ plus surfaces
- Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
- Guide ridges for perfect insert fit
- Designated feeding ports for easy media changes, suitable for robotics/automation
- 10 plates per pack (individually wrapped with lid)
Surface Information:
- pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
- cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
- Ideal for co-culture, secretions studies, and migrations studies
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Specifications
| Closure Included | Yes |
| Bottom Shape | Flat |
| Capacity (Metric) | 3.1 mL |
| Color | Transparent |
| Culture Area | 210 mm2 |
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | pureGrade S |
| Type | Cell Culture Insert System |
| Well Count | 24 |
| Show More |