Learn More
Brady™ Bradylink™ Warning Post and Chain
Product Code. 11761356 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Yellow
Quantity:
1 Ea.
6 Pk.
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
11761356 Yellow 6 Pk. Pack of 6
11721955 Black 1 Ea. Each
11741356 Black 6 Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11761356 Supplier Brady™ Supplier No. 092121
  • ONLINE EXCLUSIVE PRICE
    865.00€
    845.87€ / paquete de 6
    Ahorro 19.13€
    2% Descuento
Fecha estimada de envoi: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
This Bradylink™ Warning Post and Chain is made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.
  • Posts are non-conductive so can be safely used in electrically hazardous areas.
  • Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
  • Hollow base can be filled with sand for more stable application.
  • Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
  • Bases and tops are included.
Specifications
Product Type Warning Post and Chain
Alternate Part Number BRADYLINK WARNING POST SOLID YELLOW-PAC
Color Yellow
Material Polyethylene
Description Bradylink Warning Posts
Diameter (English) 3 in.
For Use With (Application) Traffic Control
Height (English) 41 in.
Height (Metric) 1041.4 mm
Includes Bases and tops
Quantity 6 Pk.