Learn More
Brady™ Thermal Transfer Printable Nylon Cloth Labels
Product Code. 30041427 Shop All Brady Products
Change view
Click to view available options
Size:
0.25 x 2 in. (L x W)
0.375 x 1 in. (L x W)
0.375 x 1 x 0.375 in. (L x W x dia.)
0.5 in. (dia.)
0.5 x 0.9 in. (L x W)
0.75 x 1.5 in. (L x W)
2 x 1 in. (L x W)
Quantity:
1500 Pk.
3000 Pk.
Unit Size:
1 roll
Pack of 1500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Quantity Unit Size
30041427 0.5 in. (dia.) 1500 Pk. Pack of 1500
10180213 0.375 x 1 x 0.375 in. (L x W x dia.) 3000 Pk. 1 roll
12869835 0.5 x 0.9 in. (L x W) 1500 Pk. 1 roll
15132243 2 x 1 in. (L x W) 1500 Pk. 1 roll
12624585 0.75 x 1.5 in. (L x W) 1500 Pk. 1 roll
15482143 0.375 x 1 in. (L x W) 1500 Pk. 1 roll
13089292 0.25 x 2 in. (L x W) 1500 Pk. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 30041427 Supplier Brady™ Supplier No. 116384
  • ONLINE EXCLUSIVE PRICE
    246.00€
    235.95€ / paquete de 1500
    Ahorro 10.05€
    4% Descuento
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Thermal Transfer Printable Nylon Cloth Labels with high adhesion for use in autoclave, freezer or liquid nitrogen.
  • White, matte finished labels suitable for use in laboratory identifications and vial labeling.
  • Low profile.
  • Recommended for use with Brady thermal transfer printer ribbon series 4900.
  • Compatible with color Brady series 6200, 6000, 4300, and 4500.
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL, CSA C22.2 No.0.15-95, RoHS 2002/95/EC, DIN VDE 0472 part 815
Description Thermal Transfer Printer Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Nylon Cloth
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For wiremarking and general labeling applications
For Use With (Equipment) BBP11, BBP12, THT 25 mm Core Printers
Horizontal Repeat (Metric) 15.24 mm
Includes Roll of 1500 Labels
Labels per Row 3
Life Span 2 Years
Min. Operating Temperature (Metric) -40°C
Quantity 1500 Pk.
Shape Round
Size 0.5 in. (dia.)
Size (English) 0.5 in. dia.
Size (Metric) 1.3 cm dia.
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Tensile Strength 80 lb./in.
Thickness (English) 0.0065 in.
Thickness (Metric) 0.166 mm
Vertical Repeat (Metric) 15.88 mm
Dimensions (L x W) 12.70 mm (Dia)
Length (English) 0.5 in.
Width (English) 0.5 in.
Width (Metric) 12.70 mm
Width (Metric) Printable Area 48.26 mm
Diameter (English) 0.5 in.
Diameter (Metric) 12.70 mm
Alternate Part Number THT-144-499-1.5-SC