Learn More
Thermo Scientific™ Bovine Genotypes Panel 3.1 Kit
Bovine Genotypes Panel 3.1 Kit
Marca:  Thermo Scientific™ F900L
Detalles adicionales : Peso : 0.25000kg
 Ver más versiones de este producto
Código de producto. 15451134
  • 6350.00€ / 1 set
Fecha de envío estimada
del proveedor 02-05-2019
Añadir a la cesta
Descripción
Descripción
Thermo Scientific Bovine Genotypes Panel 3.1 Kit is offered for cattle parentage testing and identification using Short Tandem Repeat (STR) DNA loci.
The Bovine Genotypes Panel 3.1 allows co-amplification of the 18 microsatellites in a single multiplex PCR reaction. One primer from each primer pair is end-labeled with a fluorescent dye. After PCR, the fragments are separated and detected in a single electrophoresis injection, using an automated electrophoresis instrument (see Figure 2). In addition, the kit includes bovine control DNA for verification of acceptable PCR and electrophoresis conditions. Reagents and protocols of the Bovine Genotypes Panel 3.1 have been optimized to deliver similar peak sizes (amplification yields) for each loci. Allele callings obtained with this kit represent the true alleles of an individual, instead of 'plus-A peaks' or 'split peaks' typically interpreted when using e.g. a conventional Taq DNA polymerase. The kit employs Phusion Hot Start DNA Polymerase that has a proofreading activity and eliminates the 'plus-A peaks'.
Thermo Scientific Bovine Genotypes Panel 3.1 encompasses all the 12 STR loci recommended by the International Society for Animal Genetics (ISAG) for routine use in parentage testing and identification, including TGLA227, BM2113, TGLA53, ETH10, SPS115, TGLA126, TGLA122, INRA23, ETH3, ETH225, BM1824, and BM1818. In addition, the kit includes the following six microsatellites, which are among the list of loci recommended by the Food and Agriculture Organization of the United Nations (FAO) for genetic studies of domestic animals: SPS113, RM067, CSRM60, MGTG4B, CSSM66, and ILSTS006. Bovine Genotypes Panel 3.1 is a combination of Panels 1.2 and 2.2, thus enabling the analysis of both panels in a single reaction.
Highlights
  • Various kit choices that include STR loci recommended by ISAG
  • Single multiplex reaction - less pipetting steps
  • Optimized reagents
  • Control DNA provided for verification of PCR and electrophoresis conditions
  • Allele calling represents true alleles (no plus-A peaks)
    This product has been developed in collaboration with Maxxam Analytics Inc.
  • Especificación
    Especificación
    Bovine Genotypes Panel 3.1 Kit, 500 rxns, Capillary Electrophoresis Detection Method, Bovine Genotyping and Parentage Testing, STR (Short Tandem Repeat) Analysis, Store at –20°C, Used For Animal Health, PCR Applications
    Animal health, PCR and real-time PCR, PCR genotyping, industrial and applied science
    Bovine Genotypes Panel 3.1 Kit
    500 rxn(s)
    Capillary electrophoresis
    5 x 1.1mL Bovine genotypes master mix, 5 x 1.1mL bovine genotypes panel 3.1 primer mix, 150μL bovine genotypes control DNA001
    500 rxn(s)
    –20°C
    Videos
    For Research Use Only. Not for use in diagnostic procedures.