Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Product Code. 16892090 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
25 mL
50 mL
100 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16892090 100 mL Each
16852090 2.5 mL Each
16862090 5 mL Each
16872090 25 mL Each
16882090 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16892090 Supplier Corning™ Supplier No. 6850
  • ONLINE EXCLUSIVE PRICE
    113.00€
    104.97€ / paquete de 1
    Ahorro 8.03€
    7% Descuento
Fecha estimada de envoi: 09-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Outer housing for BTD and dispensing reagents
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 100 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters