missing translation for 'onlineSavingsMsg'
Learn More

Boro, solución estándar de AAS, Specpure™, B 1000 μg/ml, Thermo Scientific Chemicals

Catalog Number p-7080398
Click to view available options
Cantidad:
100 mL
500 mL
Este artículo no se puede devolver. Vea la política de devoluciones
H3BO3 in H2O
MFCD00134034
Este artículo no se puede devolver. Vea la política de devoluciones
H3BO3 in H2O
MFCD00134034

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Boron, AAS standard solution
Nombre de nota B 1000μg/mL
Cantidad 100 mL
Grado Specpure™
Forma física Líquida

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'