Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
Marca:  Thermo Scientific™ C5000-72R
Detalles adicionales : Peso : 0.03000kg
 Ver más versiones de este producto
Código de producto. 15318036
  • 40.25€ / paquete de 100
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
  • AVCS closures seal as well as crimp top vials.
  • AVCS design allows for consistent sealing, eliminating user-to-user variance.
Especificaciones
Especificaciones
100/Pk
AVCS
9 mm
PTFE/Silicone
Red
Yes
Videos