missing translation for 'onlineSavingsMsg'
Learn More

BOC-DL-Valina, 98 %, Thermo Scientific Chemicals

Catalog Number p-4089982
Click to view available options
Cantidad:
10 g
50 g
Envase:
Frasco de vidrio
Este artículo no se puede devolver. Vea la política de devoluciones
54895-12-4
C10H19NO4
217.27
MFCD00069973
SZXBQTSZISFIAO-UHFFFAOYSA-N
boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine
270655
2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
CC(C)C(NC(=O)OC(C)(C)C)C(O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
54895-12-4
C10H19NO4
217.27
MFCD00069973
SZXBQTSZISFIAO-UHFFFAOYSA-N
boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine
270655
2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
CC(C)C(NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material BOC-DL-Valine
Punto de fusión 112°C to 114°C
Intervalo de porcentaje de ensayo 97.5% min. (HPLC)
Envase Frasco de vidrio
Fórmula lineal (CH3)2CHCH(COOH)NHCOOC(CH3)3
Cantidad 10 g
Formula Weight (peso de la fórmula) 217.26
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'