missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17769-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.Especificaciones
BMP9 | |
Polyclonal | |
Unconjugated | |
GDF2 | |
BMP 9, BMP9, Bone morphogenetic protein 9, GDF 2, GDF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2658 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UK05 | |
GDF2 | |
BMP9 Fusion Protein Ag12177 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |