missing translation for 'onlineSavingsMsg'
Learn More

BMP-7 Antibody (4E7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-04259AF405

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216062

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

BMP-7
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
BMP7
Recombinant human BMP7 (293-431 aa) purified from E. coli
0.1 mL
Biologically Active Proteins, Cytokine Research
655
Human
Purified
Western Blot, ELISA
40000000
Western Blot, ELISA
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1
Mouse
Protein G purified
RUO
Primary
Anti-human BMP7 mAb, clone 4E7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BMP7 protein.
Store at 4C in the dark.
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales