Learn More
Bmf Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NBP1-76666
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214397
  • 481.63€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
Bmf Polyclonal specifically detects Bmf in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
Bmf
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Rabbit
25 kDa
0.1 mg
Apoptosis, Cancer, Tumor Suppressors
90427
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 2.5-5 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin
NP_277038
BMF
Antibody was raised with a synthetic peptide corresponding to 14 amino acids near the carboxy terminus of human Bmf. The immunogen is located within the last 50 amino acids of Bmf. Amino Acid Squence: ALNGEENRNGAGPR
Affinity Purified