Learn More
Euro Protection™ Blue SPP Coverall
Product Code. 12033226
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12033226 X-Large Pack of 50
12043226 2X-Large Pack of 50
15852555 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12033226 Supplier Euro Protection™ Supplier No. 42204
  • ONLINE EXCLUSIVE PRICE
    238.19€
    82.39€ / paquete de 50
    Ahorro 155.80€
    65% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Euro Protection™ Blue SPP Coverall is lightweight, disposable and features elastic at various points for good fit.
  • Hood with tightening elastic.
  • Zipper.
  • Tightening elastic on cuffs, waist, and ankles.
  • Light weight
  • Disposable
  • Comfortable
Specifications
Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Resistance Abrasion
Certifications/Compliance (Europe) 1. Annexe II, Book III, Title I, Fourth Part of Code du Travail2. Article R412-233. EN530Article R4311-12
For Use With (Application) Visitors, Agrobusiness Industries, Cleaning, Packaging, Maintenance
Weight (Metric) 50 g/m2
Quantity 50/Pk.