Learn More
Scotch-Brite™ Blue Floor Pads
Product Code. 17198830
Change view
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17198830 460 mm Pack of 5
17118830 254 mm Pack of 5
17128830 280 mm Pack of 5
17118840 505 mm Pack of 5
17128840 530 mm Pack of 5
17138830 305 mm Pack of 5
17148830 330 mm Pack of 5
17158830 355 mm Pack of 5
17168830 380 mm Pack of 5
17178830 406 mm Pack of 5
17188830 432 mm Pack of 5
17108840 480 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 17198830 Supplier Scotch-Brite™ Supplier No. 7100182694
  • ONLINE EXCLUSIVE PRICE
    161.00€
    93.86€ / paquete de 5
    Ahorro 67.14€
    41% Descuento
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Designed for frequent burnishing with less dust and burning, used to restore the gloss to dull floors. Can be used wet for regular maintenance of unprotected floors. These pads are also recommended for dry stripping soft floors.
  • Heavy-duty scrub pad removes dirt, spills, scuffs, and 1-2 coats of finish
  • Leaves surfaces clean for recoating
  • Designed for frequent burnishing with less dust and burning
  • Washable and reusable pad is economical and helps postpone end-of-life disposal to landfills
  • Versatile pads can be used for heavy-duty spray cleaning and foam scrubbing
  • Available in sizes for rotary and automatic floor machines running at low speed (175-600 rpm)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stripping Pads
Color Blue
Diameter (Metric) 460 mm
For Use With (Application) Spray Cleaning, and Foam Scrubbing
For Use With (Equipment) Rotary and Automatic Floor Machines
Style Round