Learn More
Karl Hecht™ Blood Smear Slides
Product Code. 10544222
Change view
Click to view available options
Product Type:
Undritz blood smear slide
Unit Size:
Each
Product Code. Product Type Unit Size
10544222 Undritz blood smear slide Each
1 options
This item is not returnable. View return policy
Product Code. 10544222 Supplier Karl Hecht™ Supplier No. 40619010
  • ONLINE EXCLUSIVE PRICE
    7.46€
    3.24€ / paquete de 1
    Ahorro 4.22€
    56% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
An excellent accessory for the microscopic examination of blood samples.
  • According to Undritz
  • CE marking
  • Clear glass
  • Both sides polished
  • Beveled edges and corners
  • Thickness from 1.6 to 1.7mm
  • Dimensions approx. 40 x 20mm
Specifications
Product Type Undritz blood smear slide
For Use With (Application) Microscopic blood examination