Learn More
BIX, TRC
Product Code. 30345226
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30345226 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30345226 Supplier Toronto Research Chemicals Supplier No. TRCB591768100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material BIX
CAS 101714-41-4
Molecular Formula C9 H7 N O3 S
Quantity 100 mg
Synonym BIX, 2-(3,4-Dihydroxyphenyl)-2-oxoethyl Ester Thiocyanic Acid
InChI Formula InChI=1S/C9H7NO3S/c10-5-14-4-9(13)6-1-2-7(11)8(12)3-6/h1-3,11-12H,4H2
SMILES Oc1ccc(cc1O)C(=O)CSC#N
IUPAC Name [2-(3,4-dihydroxyphenyl)-2-oxoethyl] thiocyanate
Molecular Weight (g/mol) 209.222
Formula Weight 209.015