missing translation for 'onlineSavingsMsg'
Learn More

Sulfona de bis(4-nitrofenilo), 95 %, Thermo Scientific™

Catalog Number p-7025740
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
1156-50-9
C12H8N2O6S
308.264
MFCD00010447
BVHNGWRPAFKGFP-UHFFFAOYSA-N
bis 4-nitrophenyl sulfone, 4-nitrophenyl sulfone, 4,4'-dinitrodiphenyl sulfone, bis 4-nitrophenyl sulphone, bis p-nitrophenyl sulfone, 4,4'-dinitrodiphenylsulfone, 1,1'-sulfonylbis 4-nitrobenzene, sulfone, bis p-nitrophenyl, benzene, 1,1'-sulfonylbis 4-nitro-9ci
70871
1-Nitro-4-(4-nitrofenil)sulfonilbenceno
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
1156-50-9
C12H8N2O6S
308.264
MFCD00010447
BVHNGWRPAFKGFP-UHFFFAOYSA-N
bis 4-nitrophenyl sulfone, 4-nitrophenyl sulfone, 4,4'-dinitrodiphenyl sulfone, bis 4-nitrophenyl sulphone, bis p-nitrophenyl sulfone, 4,4'-dinitrodiphenylsulfone, 1,1'-sulfonylbis 4-nitrobenzene, sulfone, bis p-nitrophenyl, benzene, 1,1'-sulfonylbis 4-nitro-9ci
70871
1-Nitro-4-(4-nitrofenil)sulfonilbenceno
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Bis(4-nitrophenyl) sulfone
Punto de fusión 252°C to 255°C
Olor Odorless
Cantidad 5 g
Formula Weight (peso de la fórmula) 308.27
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'