Learn More
ACROBiosystems Biotinylated Human PD-L1 / B7-H1 (19-134) Protein, His,Avitag™
Recommended for biopanning
423.38€ - 1862.87€
Especificaciones
Número de acceso | NP_054862 |
---|---|
Peso molecular | 16.9 kDa |
Fuente | Biotinylated Human PD-L1 (19-134), His,Avitag™ (recommended for biopanning) (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1). |
Inmunógeno | Phe 19 - Tyr 134 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16477959
|
ACROBiosystems
PDL-H82E4-200UG |
200 μg |
1862.87€
200 microgramos |
Fecha estimada de envío: 30-04-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16487959
|
ACROBiosystems
PDL-H82E4-25UG |
25 μg |
423.38€
25 microgramos |
Fecha estimada de envío: 30-04-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 19 kDa and 21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_054862 | |
Biotinylated Human PD-L1 (19-134), His,Avitag™ (recommended for biopanning) (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1). | |
-20°C | |
PD-L1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
16.9 kDa | |
Phe 19 - Tyr 134 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |