Learn More
ACROBiosystems Biotinylated Human IL-22 Protein, His, Avitag™
Biotinylated Protein
423.38€ - 1862.87€
Especificaciones
Peso molecular | 20.3 kDa |
---|---|
Fuente | Biotinylated Human IL-22, His,Avitag™ (IL2-H8247) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZ x 6-1). |
Inmunógeno | Ala 34 - Ile 179 |
Requisitos de almacenamiento | -20°C |
Alias de gen | IL-22,IL-TIF,ZCYTO18 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17196401
|
ACROBiosystems
IL2-H8247-200UG |
200 μg |
1862.87€
200 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17106411
|
ACROBiosystems
IL2-H8247-25UG |
25 μg |
423.38€
25 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Interleukin-22 (IL22) is also known as cytokine Zcyto18, IL-10-related T-cell-derived-inducible factor (IL-TIF), which is belongs to the IL-10 family or IL-10 superfamily (including IL-19, IL-20, IL-24, and IL-26), a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells and initiates innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 biological activity is initiated by binding to a cell-surface complex composed of IL-22R1 and IL-10R2 receptor chains and further regulated by interactions with a soluble binding protein IL-22BP. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10.
This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.3 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
20.3 kDa | |
Ala 34 - Ile 179 | |
IL-22,IL-TIF,ZCYTO18 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Biotinylated Human IL-22, His,Avitag™ (IL2-H8247) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZ x 6-1). | |
-20°C | |
IL-22 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |