Learn More
ACROBiosystems Biotinylated Human CEACAM-5 / CD66e Protein, His, Avitag™
Biotinylated Protein
Marca: ACROBiosystems CE5-H82E0-25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 75.0 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_004354.3 | |
25 μg | |
Lys 35 - Ala 685 | |
CEACAM-5,CD66e,CEA,Meconium antigen 100 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
75.0 kDa | |
Biotinylated Human CEACAM-5, His,Avitag (CE5-H82E0) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). | |
-20°C | |
CEACAM-5 | |
Human | |
20 mM Tris, 150 mM Potassium glutamate, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |