Learn More
ACROBiosystems Biotinylated Human CD229 / SLAMF3 Protein, His, Avitag™
Biotinylated Protein
Marca:  ACROBiosystems CD9-H82E6-25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17196161
  • 423.38€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock
Descripción
Descripción
CD229, also known as Ly9 and SLAMF3, is a 120 kDa type I transmembrane glycoprotein in the SLAM subgroup of the CD2 family. Signaling lymphocyte activation molecule (SLAM) family receptors are critically involved in modulating innate and adaptive immune responses. CD229 is expressed on T and B cells, thymocytes, and more weakly on NK cells. Homophilic binding between CD229 molecules is mediated by the N-terminal Ig-like domain.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 48.5 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_002339.2
25 μg
Lys 48 - Lys 454
SLAMF3,LY9,Ly-9,hly9,Ly9
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
48.5 kDa
Biotinylated Human CD229, His,Avitag (CD9-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).
-20°C
CD229
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins