Learn More
ACROBiosystems Biotinylated Human BTN1A1 / Butyrophilin Protein, His, Avitag™
Biotinylated Protein
Marca: ACROBiosystems BT1-H82E6-25UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Butyrophilin subfamily 1 member A1 (BTN1A1) is also known as BTN, which is a member of the immunoglobulin superfamily and the major protein associated with fat droplets in the milk. BTN1A1 may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families. Furthermore, BTN1A1 regulates the amount of lipids and size of droplets expressed in milk and inhibits the proliferation of CD4 and CD8 T-cells activated by anti-CD3 antibodies, T-cell metabolism and IL2 and IFNG secretion.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.6 kDa. The protein migrates as 36-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001723.2 | |
27.6 kDa | |
25 µg | |
Ala 27 - Arg 242 | |
BTN1A1,Butyrophilin,BTN | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>90% as determined by SDS-PAGE | |
Biotinylated Human BTN1A1, His,Avitag (BT1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # Q13410-1). | |
-20°C | |
BTN1A1 | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |