missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody (S11B), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-81064B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213729

  • 588.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

beta Tubulin Monoclonal specifically detects beta Tubulin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

beta Tubulin
Monoclonal
Biotin
PBS
TUBB
Human MBP (microtubule-binding protein).
0.1 mL
Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
203068
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
S11B
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Rabbit
Protein A purified
RUO
Primary
Binds specifically to human beta Tubulin (no crossreactivity with alpha-tubulin).
Store at 4°C in the dark.
IgG λ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales