Learn More
beta-Actin Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NB600-503UV-0.05ML
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213665
  • 577.74€ / 0.05 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
beta-Actin
Polyclonal
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Rabbit
Affinity Purified
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Human, Mouse, Rat, Porcine, Avian, Chinese Hamster, Fish, Primate, Rabbit
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 350
50 mM Sodium Borate
ACTB
This beta-Actin Antibody was made from a synthetic peptide made to an N-terminal region of human Beta Actin. [UniProt P60709]
0.05 mL
60
Store at 4°C in the dark.