missing translation for 'onlineSavingsMsg'
Learn More

Tribromuro de benciltrimetilamonio, 98 %, Thermo Scientific Chemicals

Catalog Number 11445937
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
111865-47-5
C10H17Br3N+
390.965
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
bencil(trimetil)azanio;bromo molecular;hidrobromuro
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
Este artículo no se puede devolver. Vea la política de devoluciones
111865-47-5
C10H17Br3N+
390.965
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
bencil(trimetil)azanio;bromo molecular;hidrobromuro
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Benzyltrimethylammonium tribromide
Punto de fusión 99°C to 101°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 389.97
Porcentaje de pureza 98%

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'