missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium chloride, 97%

Product Code. 11402778
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402778 500 g 500g
11492768 100 g 100g
11412778 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402778 Supplier Thermo Scientific Alfa Aesar Supplier No. A16759.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-93-9 | C10H16ClN | 185.70 g/mol

Benzyltrimethylammonium chloride are Ccommercially important catalyst. Used in antistatic agent, detergent Sanitisers, softner for textiles and paper products, phase transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltrimethylammonium chloride are Ccommercially important catalyst. Used in antistatic agent, detergent Sanitisers, softner for textiles and paper products, phase transfer catalyst.

Solubility
Soluble in water, ethanol, and butanol and slightly soluble in butyl phthalate

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-93-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point ∼238°C (decomposition)
pH 6 to 8
Odor Odorless
Refractive Index 1.444
Quantity 500 g
Beilstein 3917255
Sensitivity Hygroscopic
Solubility Information Soluble in water,ethanol,and butanol and slightly soluble in butyl phthalate
Formula Weight 185.7
Percent Purity 97%
Chemical Name or Material Benzyltrimethylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.