missing translation for 'onlineSavingsMsg'
Learn More

Butirato de bencilo, 98 %, Thermo Scientific™

Catalog Number p-7027466
Click to view available options
Cantidad:
50 g
250 g
1000 g
Este artículo no se puede devolver. Vea la política de devoluciones
103-37-7
C11H14O2
178.231
MFCD00027133
VONGZNXBKCOUHB-UHFFFAOYSA-N
benzyl butyrate, benzyl n-butyrate, butanoic acid, phenylmethyl ester, phenylmethyl butanoate, benzyl n-butanoate, butyric acid, benzyl ester, phenylmethyl butyrate, benzylester kyseliny maselne, benzylbutyrate, benzyl butyrate natural
7650
butanoato de bencilo
CCCC(=O)OCC1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
103-37-7
C11H14O2
178.231
MFCD00027133
VONGZNXBKCOUHB-UHFFFAOYSA-N
benzyl butyrate, benzyl n-butyrate, butanoic acid, phenylmethyl ester, phenylmethyl butanoate, benzyl n-butanoate, butyric acid, benzyl ester, phenylmethyl butyrate, benzylester kyseliny maselne, benzylbutyrate, benzyl butyrate natural
7650
butanoato de bencilo
CCCC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Benzyl butyrate
Densidad 1.009
Punto de ebullición 240°C
Temperatura de inflamación 107°C (224°F)
Índice de refracción 1.493
Cantidad 50 g
Beilstein 2047625
Formula Weight (peso de la fórmula) 178.23
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'