Learn More
Benzyl Acetoacetate, TRC
Product Code. 30337823
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30337823 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30337823 Supplier Toronto Research Chemicals Supplier No. TRCB2100205G
 Restricted Item
Fecha estimada de envoi: 21-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzyl Acetoacetate
CAS 5396-89-4
Molecular Formula C11H12O3
Quantity 5 g
Synonym 3-Oxobutanoic Acid Benzyl Ester, 3-Oxobutanoic Acid Phenylmethyl Ester, 3-Oxobutyric Acid Benzyl Ester, Benzyl 3-Oxobutanoate, Benzyl 3-Oxobutyrate, Benzyl Acetoacetate, Benzyl Acetylacetate, NSC 4391, Phenylmethyl 3-Oxobutanoate
InChI Formula InChI=1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3
SMILES CC(=O)CC(=O)OCc1ccccc1
IUPAC Name benzyl 3-oxobutanoate
Molecular Weight (g/mol) 192.21
Formula Weight 192.0786