missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-ylacetic acid, 97%, Thermo Scientific™

Product Code. 10366284
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10366284 5 g 5g
10650895 250 mg 250mg
10422643 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366284 Supplier Thermo Scientific Maybridge Supplier No. CC40801DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 64175-51-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
InChI Key QWMVFCMIUUHJDH-UHFFFAOYSA-N
Synonym 2-benzofuran-3-yl acetic acid, 3-benzofuranacetic acid, benzo b furan-3-ylacetic acid, 2-1-benzofuran-3-yl acetic acid, benzofuran-3-yl-acetic acid, benzofuran-3-acetic acid, 1-benzofuran-3-ylacetic acid, 2-3-benzofuranyl acetic acid, pubchem7012, benzofuran-3-ylacetic acid
PubChem CID 2762940
IUPAC Name 2-(1-benzofuran-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=CO2)CC(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-3-ylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.