missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de benzhidrilamina, soportado por polímeros, 1 % de entrecruzamiento, 100-200 mallas, 0,5-1,5 mmol/g en PS-DVB, Thermo Scientific™

Catalog Number p-7059353
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
5267-34-5
C13H14ClN
219.712
MFCD03456188
CIHWJRSPVJBHGT-UHFFFAOYSA-N
benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
78921
difenilmetanamina; clorhidrato
C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl
Este artículo no se puede devolver. Vea la política de devoluciones
5267-34-5
C13H14ClN
219.712
MFCD03456188
CIHWJRSPVJBHGT-UHFFFAOYSA-N
benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
78921
difenilmetanamina; clorhidrato
C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Benzhydrylamine hydrochloride
Nombre de nota Polymer-supported, 1% cross-linked, 0.5 to 1.5 mmol/g on PS-DVB
Tamaño de malla 100 to 200 Mesh
Cantidad 5 g
Forma física Polvo
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'