Learn More
Bel-Art™ SP Scienceware™ Replacement Chucks for Pipette Pump™ Pipettors
Grips pipets securely. Bel-Art™ SP Scienceware™ Replacement Chucks are silicone rubber grips used with Bel-Art™ Pipette Pumps for safe, accurate and trouble-free pipetting with simple, one-handed operation.
Marca: Bel-Art™ H93789-0070
Detalles adicionales : Peso : 0.50000kg
Descripción
- For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
- Pipettes fit smoothly into these flexible tapered chucks
- Replacement chucks to fit various sizes/colors of Pipette Pumps in case of loss or damage
- Silicone rubber is resistant to acids and alkalis
- Disassemble parts for cleaning; chuck should not be autoclaved
Especificación
No | |
2mL pump pipettor | |
Silicone Rubber | |
Replacement Chucks |